Learn More
Cayman Chemical R406, 841290-80-0, 10 mg
SDP
Supplier:  Cayman Chemical 1142210
Encompass_Preferred
A potent orally available inhibitor of Syk (Ki = 30 nM); blocks FcεRI-dependent mast cell activation (EC50 = 43 nM) and, at 3 μM, reduces the release of IL-10, -12, and -13 by immune complex-pulsed dendritic cells
Catalog No. NC1390863