Learn More
Medchemexpress LLC HY-12067 10mg , R406 CAS:841290-81-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1206710MG
Encompass_Preferred
HY-12067 10mg R406 CAS: 841290-81-1 R406 is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 reduces immune complex-mediated inflammation. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9650