Learn More
Medchemexpress LLC HY-12067 5mg Medchemexpress, R406 CAS:841290-81-1 Purity:>98%
SDP
Catalog No. 502029888
Encompass_Preferred
Medchemexpress, HY-12067 5mg R406 CAS:841290-81-1 R406 is a competitive Syk inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9888 Supplier Medchemexpress LLC Supplier No. HY120675MG
Medchemexpress, HY-12067 5mg R406 CAS:841290-81-1 R406 is a competitive Syk inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.