Learn More
Medchemexpress LLC HY-15150 100mg Medchemexpress, R428 BGB324;Bemcentinib CAS:1037624-75-1 Purity:98%
SDP
Catalog No. 501638258
Encompass_Preferred
Medchemexpress,HY-15150 100mg R428 BGB324;Bemcentinib CAS:1037624-75-1 Formula:C30H34N8 R428 is a potent and selective inhibitor of Axl with an IC50 of 14 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8258 Supplier Medchemexpress LLC Supplier No. HY15150100MG
Medchemexpress,HY-15150 100mg R428 BGB324;Bemcentinib CAS:1037624-75-1 Formula:C30H34N8 R428 is a potent and selective inhibitor of Axl with an IC50 of 14 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.