Learn More
Medchemexpress LLC Bemcentinib, 1037624-75-1, MFCD21608463, 5 mg
SDP
Catalog No. NC0686481
Encompass_Preferred
Molecular Formula: C30 H34 N8, Molecular Weight: 506.655, 99.63%, Synonyms: R428; BGB324.
Catalog No. NC0686481 Supplier Medchemexpress LLC Supplier No. HY151505MG
Molecular Formula: C30 H34 N8, Molecular Weight: 506.655, 99.63%, Synonyms: R428; BGB324.