Learn More
Cayman Chemical (R)-4-Amino-3-hydroxybutyric Acid, Purity: ≥95%, CAS: 7013-07-2, 50 mg
SDP
Catalog No. 502991087
Encompass_Preferred
A GABA receptor modulator; binds to GABAA and GABAB receptors (IC50s = 1 and 0.35 µM, respectively); inhibits GABA uptake in rat brain synaptosomes (IC50 = 67 µM); induces currents in a patch-clamp assay using Xenopus oocytes expressing human GABAC (EC50 = 19 µM); inhibits electrical discharges in the amygdala in a cat model of N-amidinobenzamide-induced seizures at 20 mg/animal
Catalog No. 50-299-1087 Supplier Cayman Chemical Supplier No. 3153250 MG
A GABA receptor modulator; binds to GABAA and GABAB receptors (IC50s = 1 and 0.35 µM, respectively); inhibits GABA uptake in rat brain synaptosomes (IC50 = 67 µM); induces currents in a patch-clamp assay using Xenopus oocytes expressing human GABAC (EC50 = 19 µM); inhibits electrical discharges in the amygdala in a cat model of N-amidinobenzamide-induced seizures at 20 mg/animal