Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R5 peptide (acetate) | 99.0% | 2013.26 | 10 MG
SDP

Catalog No. 5000422861
Encompass_Preferred

R5 peptide (acetate) is a laboratory chemical intended for research use only and should be handled by qualified scientists in appropriately equipped facilities. It is a white to off-white solid with a molecular weight of 2013.26 and a high purity of 98.99%.

  • High purity: 98.99%
  • Molecular weight: 2013.26
  • Stable under recommended storage conditions
  • Requires sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-22861 Supplier Medchemexpress LLC Supplier No. HYP10510A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

R5 peptide (acetate) is a laboratory chemical intended for research use only and should be handled by qualified scientists in appropriately equipped facilities. It is a white to off-white solid with a molecular weight of 2013.26 and a high purity of 98.99%.

  • High purity: 98.99%
  • Molecular weight: 2013.26
  • Stable under recommended storage conditions
  • Requires sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.