Learn More
Alkali Scientific Alkali Scientific™ R5010 Rotating Tube Mixer With Three Rotisseries
SDP
Catalog No. 503029658
Encompass_Preferred
The BenchWaverª 3D Rocker combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional “undulating” motion commonly preferred for molecular biology applications.
Catalog No. 50-302-9658 Supplier Alkali Scientific Supplier No. RM5010
The BenchWaverª 3D Rocker combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional “undulating” motion commonly preferred for molecular biology applications.