Learn More
Selleck Chemical LLC R547 S2688-2mg
SDP
Catalog No. 503110184
Encompass_Preferred
R547 (Ro 4584820) is a potent ATP-competitive inhibitor of CDK1/2/4 with Ki of 2 nM/3 nM/1 nM It is less potent to CDK7 and GSK3 / while inactive to other kinases Phase 1
Catalog No. 50-311-0184 Supplier Selleck Chemical LLC Supplier No. S26882MG
R547 (Ro 4584820) is a potent ATP-competitive inhibitor of CDK1/2/4 with Ki of 2 nM/3 nM/1 nM It is less potent to CDK7 and GSK3 / while inactive to other kinases Phase 1