Learn More
Enzo Life Sciences Diacylglycerol Kinase Inhibitor I (R59 022) (5mg). CAS: 93076-89-2
SDP
Catalog No. 502009157
Encompass_Preferred
Potentiates the activity of protein kinase C (PKC) by inhibiting the diacylglycerol metabolism. Inhibits diacylglycerol kinase without affecting phosphodiesterase or phosphatidyl inositol kinase. It inhibits human platelet DAG kinase in intact cells (IC50=3.6μM) and in isolated membranes (IC50=6.7μM). Alternative name: 6-[2-(4-[(4-Fluorophenyl)phenylmethylene]-1-piperidinyl)ethyl]-7-methyl-5H-thiazolo-[3,2-a]pyrimidine-5-one. Purity: ≥99%. Appearance: Yellow powder. Solubility: Soluble in DMSO, THF or DMF. Insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9157 Supplier Enzo Life Sciences Supplier No. ALX430027M005
Potentiates the activity of protein kinase C (PKC) by inhibiting the diacylglycerol metabolism. Inhibits diacylglycerol kinase without affecting phosphodiesterase or phosphatidyl inositol kinase. It inhibits human platelet DAG kinase in intact cells (IC50=3.6μM) and in isolated membranes (IC50=6.7μM). Alternative name: 6-[2-(4-[(4-Fluorophenyl)phenylmethylene]-1-piperidinyl)ethyl]-7-methyl-5H-thiazolo-[3,2-a]pyrimidine-5-one. Purity: ≥99%. Appearance: Yellow powder. Solubility: Soluble in DMSO, THF or DMF. Insoluble in water. Long Term Storage: +4°C.