Learn More
Medchemexpress LLC HY-108355 5mg Medchemexpress, R59949 CAS:120166-69-0 Purity:>98%
SDP
Catalog No. 502080740
Encompass_Preferred
Medchemexpress, HY-108355 5mg R59949 CAS:120166-69-0 R59949 is a pan diacylglycerol kinase (DGK) inhibitor with an IC50 of 300 nM. R59949 strongly inhibits the activity of type I DGK α and γ and moderately attenuates the activity of type II DGK θ and κ. R59949 activates protein kinase C (PKC) by enhancing the levels of the endogenous ligand diacyl glycerol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0740 Supplier Medchemexpress LLC Supplier No. HY1083555MG
Medchemexpress, HY-108355 5mg R59949 CAS:120166-69-0 R59949 is a pan diacylglycerol kinase (DGK) inhibitor with an IC50 of 300 nM. R59949 strongly inhibits the activity of type I DGK α and γ and moderately attenuates the activity of type II DGK θ and κ. R59949 activates protein kinase C (PKC) by enhancing the levels of the endogenous ligand diacyl glycerol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.