Learn More
Apexbio Technology LLC R788 disodium, 10mg. Cas: 1025687-58-4 MFCD: MFCD15146370. Syk inhibitor,ATP competitive and potent
SDP
Supplier:  Apexbio Technology LLC A833210
Encompass_Preferred
R788, which is the prodrug of R406, is a potnet and selective Syk inhibitor. The spleen tyrosine kinase (Syk) plays a key role in the signaling of activating Fc receptors and the B-cell receptor (BCR). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5114