Learn More
Selleck Chemical LLC R788 (Fostamatinib) Disodium 50mg 1025687-58-4 Tamatinib Fosdium
SDP
Catalog No. 505941
Encompass_Preferred
R788 (Fostamatinib) disodium (Tamatinib Fosdium), a prodrug of the active metabolite R406, is a Syk inhibitor with IC50 of 41 nM in a cell-free assay, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-594-1 Supplier Selleck Chemical LLC Supplier No. S220650MG
R788 (Fostamatinib) disodium (Tamatinib Fosdium), a prodrug of the active metabolite R406, is a Syk inhibitor with IC50 of 41 nM in a cell-free assay, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.