Learn More
Selleck Chemical LLC Fostamatinib (R788) 5mg 901119-35-5
SDP
Supplier:  Selleck Chemical LLC S26255MG
Encompass_Preferred
Fostamatinib (R788), a prodrug of the active metabolite R406, is a Syk inhibitor with IC50 of 41 nM, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-685-5