Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Series 400 Half-Mask Respirators with R95 Filter Assembly
SDP

Catalog No. 19222463 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-463 Small/Medium
19-222-464 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-463 Supplier Dentec Safety™ Supplier No. 4SMDR5Z
Add to Cart
Add to Cart

Elastomeric half-mask respirators with R95 assembly are suitable for use with welding, dusts, and oil based particulate aerosols.

  • Low profile design 400 Comfort-Air™ Series complete with R95 assembly
  • Extended field of vision
  • Large inhalation valves allows users to inhale a larger volume of air per breath
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensures hot, exhaled air exits the mask immediately
  • Rolled sealing flange provides comfort and improves fit
  • Textured sealing area reduces risk of the mask slipping on the face to ensure a comfortable seal on a wide variety of face sizes
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Filter Type R95
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small/Medium
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Cartridges/Diskits
Model 4-SM-DR5Z
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.