Learn More
ACROBiosystems ACROBiosystems Rat NKG2A & CD94 Protein, Mouse IgG2a Fc Tag (100 μg) - NC4-R5258-100UG
Catalog No. 502370267
Encompass_Preferred
Rat NKG2A&CD94 Protein, Mouse IgG2a Fc Tag (NC4-R5258-100UG) is a recombinant fusion protein comprising rat NKG2A (Thr90-Asn231) and CD94 (Lys32-Thr179) expressed in HEK293 cells and fused to a mouse IgG2a Fc at the C-terminus. Supplied lyophilized (100 μg), the protein is verified by SEC-MALS and SDS-PAGE and is intended for biochemical and immunological assays.
  • High purity (>95% by SDS-PAGE; >90% by SEC-MALS).
  • MALS verified for native oligomeric state.
  • Low endotoxin (<1.0 EU/μg).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Includes defined sequence segments for NKG2A and CD94.
  • Store lyophilized at -20°C or lower; keep reconstituted material sterile at -70°C.
Catalog No. 50-237-0267 Supplier ACROBiosystems Supplier No. NC4R5258100UG
Rat NKG2A&CD94 Protein, Mouse IgG2a Fc Tag (NC4-R5258-100UG) is a recombinant fusion protein comprising rat NKG2A (Thr90-Asn231) and CD94 (Lys32-Thr179) expressed in HEK293 cells and fused to a mouse IgG2a Fc at the C-terminus. Supplied lyophilized (100 μg), the protein is verified by SEC-MALS and SDS-PAGE and is intended for biochemical and immunological assays.
  • High purity (>95% by SDS-PAGE; >90% by SEC-MALS).
  • MALS verified for native oligomeric state.
  • Low endotoxin (<1.0 EU/μg).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Includes defined sequence segments for NKG2A and CD94.
  • Store lyophilized at -20°C or lower; keep reconstituted material sterile at -70°C.