Learn More
Lauda Brinkmann Lp RA 24 REFRIGERATED CIRCULATOR
Catalog No. 50931213
Encompass_Preferred
Refrigerated Circulator, Alpha Series, Model RA 24, Digital control interface with simple 3 button operation, 24 L bath size, 425 Watts of cooling capacity at 20oC, 1.15 kW Heater, Temperature range of -25 to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 6 This item is non returnable*
Catalog No. 50-931-213 Supplier Lauda Brinkmann Lp Supplier No. L000652
Add to Cart
Add to Cart
Request a Quote
Refrigerated Circulator, Alpha Series, Model RA 24, Digital control interface with simple 3 button operation, 24 L bath size, 425 Watts of cooling capacity at 20oC, 1.15 kW Heater, Temperature range of -25 to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 6 This item is non returnable*