Learn More
Lauda Brinkmann Lp RA 8 REFRIGERATED CIRCULATOR
Catalog No. 50931211
Encompass_Preferred
Refrigerated Circulator, Alpha Series, Model RA 8, Digital control interface with simple 3 button operation, 8 L bath size, 225 Watts of cooling capacity at 20oC, 1.15 kW Heater, Temperature range of -25 to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 60H This item is non returnable*
Catalog No. 50-931-211 Supplier Lauda Brinkmann Lp Supplier No. L000650
Refrigerated Circulator, Alpha Series, Model RA 8, Digital control interface with simple 3 button operation, 8 L bath size, 225 Watts of cooling capacity at 20oC, 1.15 kW Heater, Temperature range of -25 to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 60H This item is non returnable*