Learn More
ATCC Recombinant Adeno-associated Virus 8 (rAAV8) Reference Standard Stock (ATCC VR-1816™)
Supplier:  ATCC VR1816
Encompass
Recombinant Adeno-associated virus 8 Reference Standard Stock (rAAV8 RSS) (ATCC VR-1816™) is a purified viral preparation developed by the AAV Reference Standard Working Group. This reference material is characterized for particle count, genome copy number, and infectivity and is used for calibration of vector standards in gene therapy research.
Catalog No. 50-238-2802