Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit anti-Goat IgG (Heavy Chain) Superclonal™ Secondary Antibody

Catalog No. pia27011
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA27011 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA27011 Supplier Invitrogen™ Supplier No. A27011
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Secondary Antibody

The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat IgG (Heavy Chain)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Rabbit
Immunogen Recombinant full-length protein.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.