Learn More
AdipoGen anti-Rab1-GTP, mAb (rec.) (ROF7) (100 µg)
SDP
Supplier:  AdipoGen AG27B0006C100
Encompass_Preferred
Recombinant Antibody. Recognizes human, mouse, rat and dog Rab1a-GTP and Rab1b-GTP. Species cross-reactivity: Dog, Human, Mouse, Rat. Clone: ROF7. Isotype: Human IgG2lambda. Applications: ICC, IP. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Rab1 (Ypt1 in yeast) is a small GTPase that plays a well-established role in mediating ER-to-Golgi protein transport in both yeast and mammalian cells. Rab1 is also involved in COPI vesicle formation and other distinct transport pathways, including ER-to-Golgi intermediate compartment (ERGIC)-to-cell periphery trafficking. Recently Rab1 has been shown to function in antimicrobial autophagy (autophagosome formation), as well as other forms of autophagy in mammalian cells in a way independent of ER-to-Golgi trafficking. anti-Rab1-GTP, monoclonal antibody (recombinant) (ROF7) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3024