Learn More
Aviva Systems Biology Corp RAB11B Antibody - C-terminal region (ARP48246_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48246_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB11B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAB11B. Alternate names: H-YPT3, MGC133246ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-59774