Learn More
Aviva Systems Biology Corp RAB11FIP2 Antibody - N-terminal region (ARP55095_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55095_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB11FIP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB11FIP2. Alternate names: KIAA0941, Rab11-FIP2, nRip11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-003