Learn More
Aviva Systems Biology Corp RAB11FIP5 Antibody - N-terminal region (ARP55260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB11FIP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RAB11FIP5. Alternate names: DKFZp434H018, GAF1, KIAA0857, RIP11, pp75ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-019