Learn More
Aviva Systems Biology Corp RAB14 Antibody - C-terminal region (AVARP13107_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13107_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAB14. Alternate names: FBP, RAB-14ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-53794