Learn More
Aviva Systems Biology Corp RAB15 Antibody - middle region (ARP55918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB15. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB15. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-097