Learn More
ABclonal Technology RAB1A Rabbit pAb
SDP
Supplier:  ABclonal Technology A146631000UL
Encompass_Preferred
This gene encodes a member of the Ras superfamily of GTPases. Members of the gene family cycle between inactive GDP-bound and active GTP-bound forms. This small GTPase controls vesicle traffic from the endoplasmic reticulum to the Golgi apparatus. Multiple alternatively spliced transcript variants have been identified for this gene which encode different protein isoforms.
Catalog No. 50-001-04665