Learn More
Aviva Systems Biology Corp RAB1B Antibody - C-terminal region (ARP64096_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64096_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB1B. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-172