Learn More
Medchemexpress LLC Rab1b protein, human (HEK293, Fc) | 97.9% | 10 UG
SDP
Catalog No. 5000291915
Encompass_Preferred
Recombinant human RAB1B protein expressed in HEK293 cells and fused to an Fc tag. Supplied as purified protein suitable for biochemical and cell-based assays; reported molecular weight ~58 kDa and reported purity 97.9%.
  • Recombinant human RAB1B expressed in HEK293 cells.
  • Fc-tagged fusion to aid stability and detection.
  • Purified protein with reported purity of 97.9%.
  • Approximate molecular weight ~58 kDa.
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-002-91915 Supplier Medchemexpress LLC Supplier No. HYP7599310UG
Recombinant human RAB1B protein expressed in HEK293 cells and fused to an Fc tag. Supplied as purified protein suitable for biochemical and cell-based assays; reported molecular weight ~58 kDa and reported purity 97.9%.
  • Recombinant human RAB1B expressed in HEK293 cells.
  • Fc-tagged fusion to aid stability and detection.
  • Purified protein with reported purity of 97.9%.
  • Approximate molecular weight ~58 kDa.
  • Suitable for biochemical and cell-based assays.