Learn More
Aviva Systems Biology Corp RAB23 Antibody - N-terminal region (ARP56870_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56870_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB23. Alternate names: DKFZp781H0695, HSPC137, MGC8900ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62020