Learn More
Aviva Systems Biology Corp RAB25 Antibody - middle region (ARP57401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB25. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB25. Alternate names: CATX-8, RAB11CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-307