Learn More
Aviva Systems Biology Corp RAB26 Antibody - N-terminal region (ARP55031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB26. Alternate names: V46133ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-61405