Learn More
Aviva Systems Biology Corp Rab27a Antibody - middle region (ARP56564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rab27a. It was validated on Western Blot. . Alternate names: 2210402C08Rik, 2410003M20Rik, 4933437C11Rik, ashApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-176