Learn More
Aviva Systems Biology Corp Rab27b Antibody - C-terminal region (ARP56565_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56565_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rab27b. It was validated on Western Blot. . Alternate names: 2310021G14Rik, B130064M09RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-177