Learn More
Aviva Systems Biology Corp RAB2A Antibody - C-terminal region (ARP61451_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61451_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB2A. It was validated on Western Blot. . Alternate names: RAB2, LHXApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-382