Learn More
Aviva Systems Biology Corp RAB2B Antibody - C-terminal region (ARP60691_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB2B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RAB2B. Alternate names: FLJ14824ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-128