Learn More
Aviva Systems Biology Corp RAB35 Antibody - middle region (ARP52268_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52268_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB35. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB35. Alternate names: H-ray, RAB1C, RAYApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-679