Learn More
Aviva Systems Biology Corp RAB37 Antibody - middle region (ARP55739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB37. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB37. Alternate names: FLJ30284, FLJ32507ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-074