Learn More
Aviva Systems Biology Corp RAB38 Antibody - N-terminal region (ARP42610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB38. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB38. Alternate names: NY-MEL-1, rrGTPbpApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58282