Learn More
Aviva Systems Biology Corp RAB39 Antibody - N-terminal region (ARP56965_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56965_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB39. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RAB39. Alternate names: RAB39ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-245