Learn More
Aviva Systems Biology Corp RAB3A Antibody - C-terminal region (AVARP13056_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13056_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAB3A. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Sheep
Catalog No. 50-014-62626