Learn More
Aviva Systems Biology Corp RAB3D Antibody - C-terminal region (AVARP13058_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13058_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB3D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAB3D. Alternate names: D2-2, GOV, RAB16, RAD3DApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62627