Learn More
Aviva Systems Biology Corp RAB3IP Antibody - N-terminal region (ARP53320_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53320_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB3IP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB3IP. Alternate names: FLJ14660, FLJ22548, MGC71495, RABIN3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60997