Learn More
Aviva Systems Biology Corp RAB40B Antibody - middle region (ARP52264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB40B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAB40B. Alternate names: FLJ42385, RAR, SEC4LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60708