Learn More
Aviva Systems Biology Corp RAB40C Antibody - N-terminal region (ARP57519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB40C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAB40C. Alternate names: RARL, RASL8CApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53880