Learn More
Enzo Life Sciences RAB5 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPGP006E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Rab5. The sequence is completely conserved in dog. UniProt ID: P20339 (human), P18066 (dog). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: ICC, IHC, WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7275