Learn More
Aviva Systems Biology Corp RAB5A Antibody - middle region (ARP56562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAB5A. Alternate names: RAB5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61906