Learn More
Aviva Systems Biology Corp Rab5a Antibody - middle region (ARP61182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rab5a. It was validated on Western Blot. . Alternate names: 2410015H04Rik, AI663973, AU021172ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-286