Learn More
Aviva Systems Biology Corp RAB5C Antibody - middle region (ARP57818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB5C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB5C. Alternate names: MGC117217, MGC138857, RAB5CL, RABL, L1880, RAB5LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-393