Learn More
AdipoGen anti-Rab6-GTP, mAb (rec.) (AA2) (100 µg)
SDP
Supplier:  AdipoGen AG27B0004C100
Encompass_Preferred
Recombinant Antibody. Recognizes human, mouse and Drosophila GTP-bound Rab6a and Rab6b and mutant Rab6Q72L. Does not detect Rab6•GDP. Species cross-reactivity: Drosophila, Human, Mouse. Clone: AA2. Isotype: Human IgG2lambda. Applications: ICC. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Rab6 is involved in protein transport. It is a regulator of membrane traffic from the Golgi apparatus towards the endoplasmic reticulum (ER).
Catalog No. 50-205-3020